6-Chloro-4-fluoronicotinaldehyde 98%

Supplier: Ambeed
Warning

A186650-100MG A186650-250MG
77164-074EA 162 USD
77164-074 77164-076
6-Chloro-4-fluoronicotinaldehyde 98%
6-Chloro-4-fluoronicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃ClFNO
MW: 159.54 g/mol
MDL Number: MFCD13189244
CAS Number: 1060809-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR