4-Nitrophthalimide 97%

Supplier: Ambeed
Warning

Synonyms: 5-Nitroisoindoline-1,3-dione

A186496-1000G A186496-100G A186496-25G A186496-500G A186496-5G
77191-820EA 154.5 USD
77191-820 77191-822 77191-824 77191-826 77191-828
4-Nitrophthalimide 97%
4-Nitrophthalimide

Precaución: for Research Use Only

Formula: C₈H₄N₂O₄
MW: 192.13 g/mol
MDL Number: MFCD00005884
CAS Number: 89-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR