(S)-2-(2-Hydroxyethyl)piperidine hydrochloride 98%

Supplier: Ambeed
Warning

A186306-100MG A186306-1G A186306-250MG
77191-814EA 8 USD
77191-814 77191-816 77191-818
(S)-2-(2-Hydroxyethyl)piperidine hydrochloride 98%
(S)-2-(2-Hydroxyethyl)piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO
MDL Number: MFCD09265049
CAS Number: 786684-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR