3-Carbethoxy-2-piperidone 98%

Supplier: Ambeed
Warning

A186006-10G A186006-1G A186006-25G A186006-5G A186006-100G
77191-804EA 23 USD
77191-804 77191-806 77191-808 77191-810 77615-572
3-Carbethoxy-2-piperidone 98%
3-Carbethoxy-2-piperidone

Precaución: for Research Use Only

Formula: C8H13NO3
MDL Number: MFCD00006038
CAS Number: 3731-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR