2-(Difluoromethyl)-5-nitropyridine 97%

Supplier: Ambeed
Warning

A185841-1G A185841-250MG A185841-100MG A185841-5G
77164-040EA 25 USD
77164-040 77164-042 77892-204 77892-205
2-(Difluoromethyl)-5-nitropyridine 97%
2-(Difluoromethyl)-5-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₄F₂N₂O₂
MDL Number: MFCD25476575
CAS Number: 1646152-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR