TATU (O-(7-aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) 98%

Supplier: Ambeed
Danger

A185595-1G A185595-25G
77191-802EA 13 USD
77191-802 77567-704
TATU (O-(7-aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) 98%
TATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)

Precaución: For research use only.

Formula: C₁₀H₁₅BF₄N₆O
MDL Number: MFCD08064301
CAS Number: 873798-09-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR