1,3-Dihydrobenzo[c]isothiazole 2,2-dioxide 97%

Supplier: Ambeed
Warning

A185260-100MG A185260-1G A185260-250MG A185260-5G
77164-024EA 66 USD
77164-024 77164-026 77164-028 77164-030
1,3-Dihydrobenzo[c]isothiazole 2,2-dioxide 97%
1,3-Dihydrobenzo[c]isothiazole 2,2-dioxide

Precaución: for Research Use Only

Formula: C₇H₇NO₂S
MDL Number: MFCD11043148
CAS Number: 111248-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR