2-Amino-4,5-bis((4-methoxybenzyl)oxy)benzoic acid 97%

Supplier: Ambeed
Warning

A185222-100MG A185222-250MG
77164-020EA 15.45 USD
77164-020 77164-022
2-Amino-4,5-bis((4-methoxybenzyl)oxy)benzoic acid 97%
2-Amino-4,5-bis((4-methoxybenzyl)oxy)benzoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₃NO₆
MW: 409.43 g/mol
MDL Number: MFCD27998185
CAS Number: 1609073-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR