N-(6,6-Dimethyl-5-(1-methylpiperidine-4-carbonyl)-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazol-3-yl)-3-methylbutanamide 98%

Supplier: Ambeed
Warning

Synonyms: PHA-793887

A184364-10MG A184364-1MG A184364-2MG A184364-50MG A184364-5MG A184364-100MG A184364-250MG
77191-782EA 45.32 USD
77191-782 77191-784 77191-786 77191-788 77191-790 77615-556 77615-558
N-(6,6-Dimethyl-5-(1-methylpiperidine-4-carbonyl)-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazol-3-yl)-3-methylbutanamide 98%
N-(6,6-Dimethyl-5-(1-methylpiperidine-4-carbonyl)-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazol-3-yl)-3-methylbutanamide

Precaución: for Research Use Only

Formula: C₁₉H₃₁N₅O₂
MW: 361.48 g/mol
MDL Number: MFCD17169992
CAS Number: 718630-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR