Tetrakis(4-bromophenyl)ethylene 97%

Supplier: Ambeed
Warning

Synonyms: 1,1,2,2-Tetrakis(4-bromophenyl)ethene

A184120-1G A184120-250MG A184120-25G A184120-5G
77163-978EA 10.3 USD
77163-978 77163-980 77163-982 77163-984
Tetrakis(4-bromophenyl)ethylene 97%
Tetrakis(4-bromophenyl)ethylene

Precaución: for Research Use Only

Formula: C₂₆H₁₆Br₄
MW: 648.03 g/mol
MDL Number: MFCD00667761
CAS Number: 61326-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR