3-(1-Methylpiperidin-4-yl)-1H-indol-5-ol 98%

Supplier: Ambeed
Warning

A184087-10MG A184087-1MG A184087-250MG A184087-50MG A184087-5MG
77163-968EA 17.51 USD
77163-968 77163-970 77163-972 77163-974 77163-976
3-(1-Methylpiperidin-4-yl)-1H-indol-5-ol 98%
3-(1-Methylpiperidin-4-yl)-1H-indol-5-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O
MW: 230.30 g/mol
MDL Number: MFCD01861184
CAS Number: 57477-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR