6-Bromo-2,7-naphthyridin-1(2H)-one 96%

Supplier: Ambeed
Warning

A184018-100MG A184018-250MG A184018-1G
77163-956EA 51 USD
77163-956 77163-958 77892-199
6-Bromo-2,7-naphthyridin-1(2H)-one 96%
6-Bromo-2,7-naphthyridin-1(2H)-one

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
MDL Number: MFCD18250876
CAS Number: 1260671-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR