5-(4-Hydroxyphenyl)-3H-1,2-dithiole-3-thione 97%
Supplier: Ambeed
|
Warning
|
A183411-100MG
A183411-10G
A183411-1G
A183411-250MG
A183411-25G
A183411-5G
77163-942EA
50.47
USD
77163-942
77163-944
77163-946
77163-948
77163-950
77163-952
5-(4-Hydroxyphenyl)-3H-1,2-dithiole-3-thione 97%
5-(4-Hydroxyphenyl)-3H-1,2-dithiole-3-thione
Precaución: for Research Use Only
Formula:
C₉H₆OS₃ MW: 226.33 g/mol |
MDL Number:
MFCD20922704 CAS Number: 18274-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...