1-(5-Chloro-2-nitrophenyl)-1-ethanone 98%

Supplier: Ambeed
Warning

Synonyms: 5'-Chloro-2'-nitroacetophenone, 1-(5-Chloro-2-nitrophenyl)ethanone

A183251-100G A183251-10G A183251-1G A183251-250MG A183251-25G A183251-5G
77191-756EA 307 USD
77191-756 77191-758 77191-760 77191-762 77191-764 77191-766
1-(5-Chloro-2-nitrophenyl)-1-ethanone 98%
1-(5-Chloro-2-nitrophenyl)-1-ethanone

Precaución: for Research Use Only

Formula: C₈H₆ClNO₃
MW: 199.59 g/mol
MDL Number: MFCD03787322
CAS Number: 18640-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR