5-Cyano-2-fluorobenzaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-3-formylbenzonitrile

A182608-10G A182608-1G A182608-250MG A182608-25G A182608-5G
77191-724EA 38 USD
77191-724 77191-726 77191-728 77191-730 77191-732
5-Cyano-2-fluorobenzaldehyde 95%
5-Cyano-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄FNO
MW: 149.1219 g/mol
MDL Number: MFCD04974126
CAS Number: 146137-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR