2-Bromo-5-chloroaniline 98%

Supplier: Ambeed
Warning

A182475-100G A182475-10G A182475-1G A182475-25G A182475-5G
77163-922EA 72.1 USD
77163-922 77163-924 77163-926 77163-928 77163-930
2-Bromo-5-chloroaniline 98%
2-Bromo-5-chloroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrClN
MDL Number: MFCD00672942
CAS Number: 823-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR