2-Cyclohexylethylamine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Aminoethyl)cyclohexane hydrochloride, 2-Cyclohexylethylamine hydrochloride
A182436-100MG
A182436-1G
77163-920EA
33
USD
77163-920
77892-192
2-Cyclohexylethylamine hydrochloride 98%
2-Cyclohexylethylamine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₈ClN |
MDL Number:
MFCD01734731 CAS Number: 5471-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...