(-)-Blebbistatin 98%

Supplier: Ambeed
Warning

Synonyms: (S)-3a-Hydroxy-6-methyl-1-phenyl-1,2,3,3a-tetrahydro-4H-pyrrolo[2,3-b]quinolin-4-one

A182285-10MG A182285-1MG A182285-5MG A182285-50MG
77191-718EA 149 USD
77191-718 77191-720 77191-722 77746-185
(-)-Blebbistatin 98%
(-)-Blebbistatin

Precaución: for Research Use Only

Formula: C₁₈H₁₆N₂O₂
MW: 292.3318 g/mol
MDL Number: MFCD08460907
CAS Number: 856925-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR