N-(9-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide 97%

Supplier: Ambeed
Warning

A182177-1G A182177-5G A182177-25G
77163-912EA 38 USD
77163-912 77163-914 77567-672
N-(9-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide 97%
N-(9-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide

Precaución: For research use only.

Formula: C₁₈H₁₉N₅O₅
MW: 385.37 g/mol
MDL Number: MFCD15145188
CAS Number: 85079-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR