3-Bromo-9-(4-bromophenyl)-9H-carbazole 97%
Supplier: Ambeed
|
Warning
|
A182073-100MG
A182073-1G
A182073-250MG
A182073-5G
77191-708EA
5.15
USD
77191-708
77191-710
77191-712
77191-714
3-Bromo-9-(4-bromophenyl)-9H-carbazole 97%
3-Bromo-9-(4-bromophenyl)-9H-carbazole
Precaución: for Research Use Only
Formula:
C₁₈H₁₁Br₂N MW: 401.10 g/mol |
MDL Number:
MFCD30063199 CAS Number: 1226860-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...