1,2-Dimethylimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dimethyl-1H-imidazole

A181857-100G A181857-25G A181857-500G
77191-696EA 11.33 USD
77191-696 77191-698 77191-700
1,2-Dimethylimidazole 98%
1,2-Dimethylimidazole

Precaución: for Research Use Only

Formula: C₅H₈N₂
MW: 96.13 g/mol
MDL Number: MFCD00005294
CAS Number: 1739-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR