2,5-Difluoro-3-nitropyridine 97%
Supplier: Ambeed
|
Warning
|
A181128-250MG
A181128-1G
A181128-100MG
77163-896EA
169.95
USD
77163-896
77593-156
77683-453
2,5-Difluoro-3-nitropyridine 97%
2,5-Difluoro-3-nitropyridine
Precaución: For research use only.
Formula:
C₅H₂F₂N₂O₂ MW: 160.08 g/mol |
MDL Number:
MFCD11519035 CAS Number: 179558-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...