3-(Dimethylamino)pyrrolidine 98%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: N,N-Dimethylpyrrolidin-3-amine

A181038-1G A181038-250MG A181038-5G A181038-10G
77163-890EA 51.5 USD
77163-890 77163-892 77163-894 77683-452
3-(Dimethylamino)pyrrolidine 98%, Ambeed.Inc
3-(Dimethylamino)pyrrolidine

Precaución: For research use only.

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD01569246
CAS Number: 69478-75-7
UN: 2734
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR