5-Amino-2-methoxypyridine-4-carboxylic Acid 96%

Supplier: Ambeed
Warning

A180846-100MG A180846-1G A180846-250MG A180846-5G
77191-672EA 50.47 USD
77191-672 77191-674 77191-676 77191-678
5-Amino-2-methoxypyridine-4-carboxylic Acid 96%
5-Amino-2-methoxypyridine-4-carboxylic Acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
MDL Number: MFCD03411719
CAS Number: 183741-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR