2-(4-Aminophenyl)benzoxazol-5-amine 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Aminophenyl)benzo[d]oxazol-5-amine, 5-Amino-2-(4-aminophenyl)benzoxazole

A180699-10G A180699-1G A180699-25G A180699-5G
77163-868EA 38.11 USD
77163-868 77163-870 77163-872 77163-874
2-(4-Aminophenyl)benzoxazol-5-amine 97%
2-(4-Aminophenyl)benzoxazol-5-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₃O
MW: 225.25 g/mol
MDL Number: MFCD00445970
CAS Number: 13676-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR