(3aR,4R,6S,6aS)-Methyl 4-((tert-butoxycarbonyl)amino)-3-(pentan-3-yl)-4,5,6,6a-tetrahydro-3aH-cyclopenta[d]isoxazole-6-carboxylate 95%

Supplier: Ambeed
Warning

A180683-100G A180683-10G A180683-1G A180683-25G A180683-5G
77163-858EA 406.85 USD
77163-858 77163-860 77163-862 77163-864 77163-866
(3aR,4R,6S,6aS)-Methyl 4-((tert-butoxycarbonyl)amino)-3-(pentan-3-yl)-4,5,6,6a-tetrahydro-3aH-cyclopenta[d]isoxazole-6-carboxylate 95%
(3aR,4R,6S,6aS)-Methyl 4-((tert-butoxycarbonyl)amino)-3-(pentan-3-yl)-4,5,6,6a-tetrahydro-3aH-cyclopenta[d]isoxazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₃₀N₂O₅
MW: 354.44 g/mol
MDL Number: MFCD18206350
CAS Number: 229613-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR