4-Hydroxy-6-methylnicotinic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Carboxy-6-methyl-4-pyridone, 6-Methyl-4-oxo-1,4-dihydropyridine-3-carboxylic acid
A179892-1000G
A179892-100G
A179892-10G
A179892-25G
A179892-500G
A179892-5G
77191-646EA
271.92
USD
77191-646
77191-648
77191-650
77191-652
77191-654
77191-656
4-Hydroxy-6-methylnicotinic acid 98%
4-Hydroxy-6-methylnicotinic acid
Precaución: for Research Use Only
Formula:
C₇H₇NO₃ MW: 153.137 g/mol |
MDL Number:
MFCD04038153 CAS Number: 33821-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...