(R)-(+)-α-Methyl-4-nitrobenzylamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (R)-α-Methyl-4-nitrobenzylamine HCl, (R)-1-(4-Nitrophenyl)ethanamine hydrochloride, (R)-4-Nitrophenyl-1-ethylamine hydrochloride

A179742-250MG A179742-5G
77191-644EA 19.57 USD
77191-644 77746-169
(R)-(+)-α-Methyl-4-nitrobenzylamine hydrochloride 97%
(R)-(+)-α-Methyl-4-nitrobenzylamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₂O₂
MW: 202.638 g/mol
MDL Number: MFCD00066311
CAS Number: 57233-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR