tert-Butyl (4-hydroxy-2-nitrophenyl)carbamate 95%

Supplier: Ambeed
Warning

A179444-100MG A179444-1G A179444-250MG A179444-5G
77163-830EA 8 USD
77163-830 77163-832 77163-834 77163-836
tert-Butyl (4-hydroxy-2-nitrophenyl)carbamate 95%
tert-Butyl (4-hydroxy-2-nitrophenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₅
MDL Number: MFCD21170216
CAS Number: 201811-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR