1-Bromo-3-chloro-2-nitrobenzene 95%

Supplier: Ambeed
Warning

A179376-10G A179376-1G A179376-250MG A179376-5G
77163-822EA 204 USD
77163-822 77163-824 77163-826 77163-828
1-Bromo-3-chloro-2-nitrobenzene 95%
1-Bromo-3-chloro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MDL Number: MFCD11100175
CAS Number: 59772-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR