2-Acetamidoisonicotinic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Acetylaminoisonicotinic acid

A179332-10G A179332-1G A179332-250MG A179332-25G A179332-5G
77163-812EA 159.65 USD
77163-812 77163-814 77163-816 77163-818 77163-820
2-Acetamidoisonicotinic acid 95%
2-Acetamidoisonicotinic acid

Precaución: for Research Use Only

Formula: C₈H₈N₂O₃
MW: 180.1600 g/mol
MDL Number: MFCD00233713
CAS Number: 54221-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR