8-Bromo-1-octene 98%

Supplier: Ambeed
Warning

A178881-100G A178881-10G A178881-25G A178881-5G A178881-1G
77163-802EA 189.52 USD
77163-802 77163-804 77163-806 77163-808 77683-433
8-Bromo-1-octene 98%
8-Bromo-1-octene

Precaución: For research use only.

Formula: C₈H₁₅Br
MW: 191.11 g/mol
MDL Number: MFCD00000275
CAS Number: 2695-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR