4-Bromo-2,6-diaminopyridine 97%

Supplier: Ambeed
Warning

A178675-250MG A178675-5G A178675-1G A178675-100MG
77163-790EA 40.17 USD
77163-790 77163-792 77550-152 77683-432
4-Bromo-2,6-diaminopyridine 97%
4-Bromo-2,6-diaminopyridine

Precaución: For research use only.

Formula: C₅H₆BrN₃
MDL Number: MFCD04115308
CAS Number: 329974-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR