2-Chloro-5,6,7,8-tetrahydro-1,6-naphthyridine 97%

Supplier: Ambeed
Warning

A178380-250MG A178380-100MG
77191-618EA 121.54 USD
77191-618 77746-161
2-Chloro-5,6,7,8-tetrahydro-1,6-naphthyridine 97%
2-Chloro-5,6,7,8-tetrahydro-1,6-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₉ClN₂
MW: 168.62 g/mol
MDL Number: MFCD10566728
CAS Number: 210539-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR