Methyl 5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylate 98%

Supplier: Ambeed
Warning

A178091-100MG A178091-1G A178091-250MG
77163-782EA 23.69 USD
77163-782 77163-784 77163-786
Methyl 5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylate 98%
Methyl 5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD28134587
CAS Number: 1454848-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR