Ethyl 4-amino-3-bromobenzoate 95%

Supplier: Ambeed
Warning

A177853-10G A177853-1G
77163-768EA 89.61 USD
77163-768 77683-425
Ethyl 4-amino-3-bromobenzoate 95%
Ethyl 4-amino-3-bromobenzoate

Precaución: For research use only.

Formula: C₉H₁₀BrNO₂
MW: 244.09 g/mol
MDL Number: MFCD03407825
CAS Number: 7149-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR