3-Methoxy-4-(trifluoromethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A176920-1G A176920-250MG A176920-5G
77163-748EA 19 USD
77163-748 77163-750 77163-752
3-Methoxy-4-(trifluoromethyl)benzoic acid 95%
3-Methoxy-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
MDL Number: MFCD13185682
CAS Number: 276861-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR