4-Chloro-3-quinolinamine 98%

Supplier: Ambeed
Warning

A176824-10G A176824-1G A176824-250MG A176824-5G A176824-25G
77163-740EA 137 USD
77163-740 77163-742 77163-744 77163-746 77892-167
4-Chloro-3-quinolinamine 98%
4-Chloro-3-quinolinamine

Precaución: for Research Use Only

Formula: C₉H₇ClN₂
MDL Number: MFCD00234511
CAS Number: 58401-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR