α,α,α,5-Tetrafluoro-o-toluic acid 98%

Supplier: Ambeed
Warning

A176699-100MG A176699-10G A176699-1G A176699-250MG A176699-5G
77191-576EA 5 USD
77191-576 77191-578 77191-580 77191-582 77191-584
α,α,α,5-Tetrafluoro-o-toluic acid 98%
α,α,α,5-Tetrafluoro-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₂
MDL Number: MFCD00083521
CAS Number: 654-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR