(2R,3S,4R)-2-(Hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol 98%

Supplier: Ambeed
Warning

A176371-100G A176371-1G A176371-25G A176371-5G
77191-562EA 349 USD
77191-562 77191-564 77191-566 77191-568
(2R,3S,4R)-2-(Hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol 98%
(2R,3S,4R)-2-(Hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol

Precaución: for Research Use Only

Formula: C₆H₁₀O₄
MDL Number: MFCD09039277
CAS Number: 13265-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR