(E)-N-Methyl-N-((3-methylbenzofuran-2-yl)methyl)-3-(7-oxo-5,6,7,8-tetrahydro-1,8-naphthyridin-3-yl)acrylamide 98%

Supplier: Ambeed
Warning

A176345-10MG A176345-1MG A176345-2MG A176345-50MG A176345-5MG
77163-726EA 140 USD
77163-726 77163-728 77163-730 77163-732 77163-734
(E)-N-Methyl-N-((3-methylbenzofuran-2-yl)methyl)-3-(7-oxo-5,6,7,8-tetrahydro-1,8-naphthyridin-3-yl)acrylamide 98%
(E)-N-Methyl-N-((3-methylbenzofuran-2-yl)methyl)-3-(7-oxo-5,6,7,8-tetrahydro-1,8-naphthyridin-3-yl)acrylamide

Precaución: for Research Use Only

Formula: C₂₂H₂₁N₃O₃
MDL Number: MFCD18633190
CAS Number: 620175-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR