Boc-Asp-OMe 97%

Supplier: Ambeed
Warning

A176233-100G A176233-10G A176233-1G A176233-25G A176233-500G A176233-5G
77163-704EA 151 USD
77163-704 77163-706 77163-708 77163-710 77163-712 77163-714
Boc-Asp-OMe 97%
Boc-Asp-OMe

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₆
MDL Number: MFCD03094777
CAS Number: 98045-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR