2-(4-Methylpiperidin-1-yl)phenylamine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(4-Methylpiperidin-1-yl)aniline
A175854-1G
A175854-250MG
77191-554EA
165.83
USD
77191-554
77191-556
2-(4-Methylpiperidin-1-yl)phenylamine 97%
2-(4-Methylpiperidin-1-yl)phenylamine
Precaución: for Research Use Only
Formula:
C₁₂H₁₈N₂ MW: 190.29 g/mol |
MDL Number:
MFCD04166381 CAS Number: 252758-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...