4,4'-Dimethyl-[1,1'-biphenyl]-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A175823-100MG A175823-1G A175823-250MG
77191-548EA 18.54 USD
77191-548 77191-550 77191-552
4,4'-Dimethyl-[1,1'-biphenyl]-2-carboxylic acid 97%
4,4'-Dimethyl-[1,1'-biphenyl]-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
MDL Number: MFCD18318953
CAS Number: 537712-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR