2-(1-(Ethylsulfonyl)azetidin-3-ylidene)acetonitrile 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A175628-1G A175628-5G A175628-25G
77163-696EA 12 USD
77163-696 77163-698 77683-411
2-(1-(Ethylsulfonyl)azetidin-3-ylidene)acetonitrile 97%, Ambeed.Inc
2-(1-(Ethylsulfonyl)azetidin-3-ylidene)acetonitrile

Precaución: For research use only.

Formula: C₇H₁₀N₂O₂S
MDL Number: MFCD22987586
CAS Number: 1187595-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR