4,4,4-Trifluoro-2-(4-fluorophenyl)-3-oxobutanenitrile 95%

Supplier: Ambeed
Warning

A175600-1G A175600-5G
77163-692EA 130 USD
77163-692 77163-694
4,4,4-Trifluoro-2-(4-fluorophenyl)-3-oxobutanenitrile 95%
4,4,4-Trifluoro-2-(4-fluorophenyl)-3-oxobutanenitrile

Precaución: for Research Use Only

Formula: C₁₀H₅F₄NO
MDL Number: MFCD09878887
CAS Number: 3108-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR