5-Chloro-2-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (5-Chloro-2-methoxyphenyl)boronic acid, 5-Chloro-2-methoxybenzeneboronic acid

A175212-10G A175212-1G A175212-25G A175212-500G A175212-5G
77191-538EA 10 USD
77191-538 77191-540 77191-542 77191-544 77191-546
5-Chloro-2-methoxyphenylboronic acid 98%
5-Chloro-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₃
MW: 186.40 g/mol
MDL Number: MFCD01318966
CAS Number: 89694-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR