1-(3,4-Dimethylphenyl)-3-methyl-1H-pyrazol-5(4H)-one 97%

Supplier: Ambeed
Warning

A174775-1G A174775-250MG
77191-526EA 8.24 USD
77191-526 77191-528
1-(3,4-Dimethylphenyl)-3-methyl-1H-pyrazol-5(4H)-one 97%
1-(3,4-Dimethylphenyl)-3-methyl-1H-pyrazol-5(4H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O
MW: 202.25 g/mol
MDL Number: MFCD00638319
CAS Number: 18048-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR