BMS-833923 98+%

Supplier: Ambeed
Warning

A174690-10MG A174690-25MG A174690-50MG
77191-516EA 30 USD
77191-516 77191-518 77191-520 77191-522 77191-524
BMS-833923 98+%
BMS-833923

Precaución: for Research Use Only

Formula: C₃₀H₂₇N₅O
MDL Number: MFCD25976660
CAS Number: 1059734-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR