(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-Cyclohexane-1,2-dicarboxylic acid, (1R,2R)-(-)-trans-Cyclohexane-1,2-dicarboxylic acid

A174004-100G A174004-10G A174004-1G A174004-25G A174004-500G A174004-5G
77191-494EA 79 USD
77191-494 77191-496 77191-498 77191-500 77191-502 77191-504
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid 98%
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.18
MDL Number: MFCD00062973
CAS Number: 46022-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR